3-Cyclohexen-1-ylbenzene
Synonym(s):
4-Phenylcyclohexene
Empirical Formula (Hill Notation):
C12H14
CAS Number:
Molecular Weight:
158.24
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing