
2-Ethylhexyl methacrylate

Synonym(s):
2-Ethylhexyl methacrylate
Linear Formula:
CH2=C(CH3)COOCH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
198.30
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-2 of 2