
1H,1H-Heptafluoro-1-butanol

Synonym(s):
2,2,3,3,4,4,4-Heptafluoro-1-butanol, Perfluoropropyl carbinol
Linear Formula:
CF3CF2CF2CH2OH
CAS Number:
Molecular Weight:
200.05
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing