![12(S)-Hydroxy-(5Z,8Z,10E,14Z)-eicosatetraenoic acid ~100 μg/mL in ethanol, ≥95% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/311/084/044d810e-c60b-41ff-a343-765e99c28855/640/044d810e-c60b-41ff-a343-765e99c28855.png)
12(S)-Hydroxy-(5Z,8Z,10E,14Z)-eicosatetraenoic acid
![12(S)-Hydroxy-(5Z,8Z,10E,14Z)-eicosatetraenoic acid ~100 μg/mL in ethanol, ≥95% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/311/084/044d810e-c60b-41ff-a343-765e99c28855/640/044d810e-c60b-41ff-a343-765e99c28855.png)
Synonym(s):
(S)-12-HETE
Empirical Formula (Hill Notation):
C20H32O3
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing