Merck
Genome sequencing and transcriptome analysis of
Biotechnology for biofuels (2017-09-16)
Christa Ivanova, Jonas Ramoni, Thiziri Aouam, Alexa Frischmann, Bernhard Seiboth, Scott E Baker, Stéphane Le Crom, Sophie Lemoine, Antoine Margeot, Frédérique Bidard
ABSTRACT
The hydrolysis of biomass to simple sugars used for the production of biofuels in biorefineries requires the action of cellulolytic enzyme mixtures. During the last 50 years, the ascomycete Sequence comparison of the cellulase-negative strain QM9978 to the reference strain QM6a identified a total of 43 mutations, of which 33 were located either close to or in coding regions. From those, we identified 23 single-nucleotide variants, nine InDels, and one translocation. The translocation occurred between chromosomes V and VII, is located upstream of the putative transcription factor We were able to establish a link between a chromosomal translocation in QM9978 and the cellulase-negative phenotype of the strain. We identified the transcription factor
Sigma-Aldrich
3-(Benzyldimethylammonio)propanesulfonate, BioXtra, ≥99.0% (HPCE)