Merck
  • RPTOR, a novel target of miR-155, elicits a fibrotic phenotype of cystic fibrosis lung epithelium by upregulating CTGF.
RPTOR, a novel target of miR-155, elicits a fibrotic phenotype of cystic fibrosis lung epithelium by upregulating CTGF.
RNA biology (2016-06-11)
Motohiro Tsuchiya, Swathi Kalurupalle, Parameet Kumar, Sarani Ghoshal, Yongqing Zhang, Elin Lehrmann, Kevin G Becker, Myriam Gorospe, Roopa Biswas
ABSTRACT
Cystic fibrosis (CF) is an autosomal recessive disorder caused by mutations in the cystic fibrosis transmembrane conductance regulator (CFTR) gene, the most frequent of which is F508del-CFTR. CF is characterized by excessive secretion of pro-inflammatory mediators into the airway lumen, inducing a highly inflammatory cellular phenotype. This process triggers fibrosis, causing airway destruction and leading to high morbidity and mortality. We previously reported that miR-155 is upregulated in CF lung epithelial cells, but the molecular mechanisms by which miR-155 affects the disease phenotype is not understood. Here we report that RPTOR (regulatory associated protein of mTOR, complex 1) is a novel target of miR-155 in CF lung epithelial cells. The suppression of RPTOR expression and subsequent activation of TGF-β signaling resulted in the induction of fibrosis by elevating connective tissue growth factor (CTGF) abundance in CF lung epithelial cells. Thus, we propose that miR-155 might regulate fibrosis of CF lungs through the increased CTGF expression, highlighting its potential value in CF therapy.
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Tablets, For General Use
Sigma-Aldrich
Anti-Raptor Antibody, Upstate®, from rabbit