Merck
  • Heliolactone, a non-sesquiterpene lactone germination stimulant for root parasitic weeds from sunflower.
Heliolactone, a non-sesquiterpene lactone germination stimulant for root parasitic weeds from sunflower.
Phytochemistry (2014-12-03)
Kotomi Ueno, Toshio Furumoto, Shuhei Umeda, Masaharu Mizutani, Hirosato Takikawa, Rossitza Batchvarova, Yukihiro Sugimoto
ABSTRACT
Root exudates of sunflower (Helianthus annuus L.) line 2607A induced germination of seeds of root parasitic weeds Striga hermonthica, Orobanche cumana, Orobanche minor, Orobanche crenata, and Phelipanche aegyptiaca. Bioassay-guided purification led to the isolation of a germination stimulant designated as heliolactone. FT-MS analysis indicated a molecular formula of C20H24O6. Detailed NMR spectroscopic studies established a methylfuranone group, a common structural component of strigolactones connected to a methyl ester of a C14 carboxylic acid via an enol ether bridge. The cyclohexenone ring is identical to that of 3-oxo-α-ionol and the other part of the molecule corresponds to an oxidized carlactone at C-19. It is a carlactone-type molecule and functions as a germination stimulant for seeds of root parasitic weeds. Heliolactone induced seed germination of the above mentioned root parasitic weeds, while dehydrocostus lactone and costunolide, sesquiterpene lactones isolated from sunflower root exudates, were effective only on O. cumana and O. minor. Heliolactone production in aquacultures increased when sunflower seedlings were grown hydroponically in tap water and decreased on supplementation of the culture with either phosphorus or nitrogen. Costunolide, on the other hand, was detected at a higher concentration in well-nourished medium as opposed to nutrient-deficient media, thus suggesting a contrasting contribution of heliolactone and the sesquiterpene lactone to the germination of O. cumana under different soil fertility levels.
Sigma-Aldrich
Isophorone, ≥97%, FG
Sigma-Aldrich
Isophorone, 97%
Sigma-Aldrich
Costunolide, ≥97% (HPLC)