Merck
  • Safety of stavudine in the treatment of HIV infection with a special focus on resource-limited settings.
Safety of stavudine in the treatment of HIV infection with a special focus on resource-limited settings.
Expert opinion on drug safety (2008-05-09)
Alain Makinson, Vincent Le Moing, Charles Kouanfack, Christian Laurent, Eric Delaporte
ABSTRACT
Western randomized trials and prospective cohorts in resource-limited settings have proven virological success with stavudine-based highly active antiretroviral therapy. However, stavudine is no longer recommended in first-line treatments in these two settings due to its intrinsic toxicities and side effects. Yet it remains a cornerstone of treatment in resource-limited settings, due to lack of alternatives and its availability in generic fixed-dose combinations. To review the toxic effects of stavudine and their prevention and management strategies, especially in resource-limited settings. Data from clinical and pharmacological trials in Western countries, as well as prospective cohorts in resource-limited settings, were reviewed. Initiating or switching to less toxic nucleoside analogues whenever possible, or lowering stavudine doses to 30 mg b.i.d., is strongly recommended.
Sigma-Aldrich
2′,3′-Didehydro-3′-deoxythymidine, ≥98% (TLC)
Stavudine, European Pharmacopoeia (EP) Reference Standard
Stavudine for system suitability, European Pharmacopoeia (EP) Reference Standard