Merck
Single- and dual-parameter FRET kinase probes based on pleckstrin.
Nature protocols (2007-04-05)
Justin Brumbaugh, Andreas Schleifenbaum, Gunter Stier, Michael Sattler, Carsten Schultz
ABSTRACT
Here we describe protocols for preparing and using fluorescent probes that respond to conformational changes by altered Foerster resonance energy transfer (FRET) efficiencies upon phosphorylation or, in principle, other posttranslational modifications (PTMs). The sensor protein, a truncated version of pleckstrin, is sandwiched between short-wavelength-excitation green fluorescent protein (GFP2) and yellow fluorescent protein (EYFP). As a result of complex conformational changes of the protein upon phosphorylation, the introduction of a second PTM consensus sequence bestows sensitivity to a second modification and yields a dual-parameter probe. The first phase of the protocol lays out the cloning strategy for single- and dual-parameter FRET sensors, including the construction of a versatile platform into which different consensus sequences may be inserted to create diverse probes. Protocols for fluorescence microscopy of the probes in living cells and image processing are also described. Probe preparation takes 7 d; microscopy and image processing take 2 h.
Sigma-Aldrich
Phorbol-12-myristate-13-acetate, Phorbol-12-myristate-13-acetate, CAS 16561-29-8, is the most common phorbol ester. Activates PKC at nanomolar concentrations.
Sigma-Aldrich
Gö 6983, A potent, cell-permeable, reversible, and ATP-competitive inhibitor of protein kinase C (PKC) that inhibits several PKC isozymes.
Sigma-Aldrich
Histamine, ≥97.0%