Merck
  • The ubiquitin ligase HOIL-1L regulates immune responses by interacting with linear ubiquitin chains.
The ubiquitin ligase HOIL-1L regulates immune responses by interacting with linear ubiquitin chains.
iScience (2021-11-11)
Carlos Gomez-Diaz, Gustav Jonsson, Katrin Schodl, Luiza Deszcz, Annika Bestehorn, Kevin Eislmayr, Jorge Almagro, Anoop Kavirayani, Mayu Seida, Lilian M Fennell, Astrid Hagelkruys, Pavel Kovarik, Josef M Penninger, Fumiyo Ikeda
ABSTRACT
The Linear Ubiquitin Chain Assembly Complex (LUBAC), composed of HOIP, HOIL-1L, and SHARPIN, promotes tumor necrosis factor (TNF)-dependent NF-κB signaling in diverse cell types. HOIL-1L contains an Npl4 Zinc Finger (NZF) domain that specifically recognizes linear ubiquitin chains, but its physiological role in vivo has remained unclear. Here, we demonstrate that the HOIL-1L NZF domain has important regulatory functions in inflammation and immune responses in mice. We generated knockin mice (Hoil-1l T201A;R208A/T201A;R208A ) expressing a HOIL-1L NZF mutant and observed attenuated responses to TNF- and LPS-induced shock, including prolonged survival, stabilized body temperature, reduced cytokine production, and liver damage markers. Cells derived from Hoil-1l T201A;R208A/T201A;R208A mice show reduced TNF-dependent NF-κB activation and incomplete recruitment of HOIL-1L into TNF Receptor (TNFR) Complex I. We further show that HOIL-1L NZF cooperates with SHARPIN to prevent TNFR-dependent skin inflammation. Collectively, our data suggest that linear ubiquitin-chain binding by HOIL-1L regulates immune responses and inflammation in vivo.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Formalin solution, neutral buffered, 10%, histological tissue fixative
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Anti-RNF31 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
GeneJuice® Transfection Reagent, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
N-Ethylmaleimide, crystalline, ≥98% (HPLC)
Sigma-Aldrich
Ubiquitin from bovine erythrocytes, BioUltra, ≥98% (SDS-PAGE), essentially salt-free, lyophilized powder
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Keratin 14 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-HOIL-1 Antibody, clone 2E2, clone 2E2, from mouse
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial