Merck
  • Effects of acid and lactone forms of statins on S-warfarin 7-hydroxylation catalyzed by human liver microsomes and recombinant CYP2C9 variants (CYP2C9.1 and CYP2C9.3).
Effects of acid and lactone forms of statins on S-warfarin 7-hydroxylation catalyzed by human liver microsomes and recombinant CYP2C9 variants (CYP2C9.1 and CYP2C9.3).
Drug metabolism and pharmacokinetics (2020-12-21)
Ayaka Shiozawa, Satoshi Yamaori, Shinobu Kamijo, Shigeru Ohmori
ABSTRACT
The inhibition of CYP2C9-mediated warfarin metabolism by acid or lactone forms of statin converted in the body and effects of CYP2C9 genetic variants on their inhibition are not fully understood. Here, the effects of acid and lactone forms of statins on S-warfarin 7-hydroxylation were investigated in vitro. S-Warfarin 7-hydroxylase activities of human liver microsomes (HLMs), recombinant CYP2C9.1 (rCYP2C9.1), and rCYP2C9.3 (Ile359Leu variant) in the presence of statins were determined by high-performance liquid chromatography. Lactone forms of atorvastatin, lovastatin, pitavastatin, pravastatin, rosuvastatin, and simvastatin inhibited the activity of HLMs more potently than the corresponding acid forms, whereas fluvastatin acid showed stronger inhibition than fluvastatin lactone. When the effects of statins on rCYP2C9 variants were examined, inhibition profiles of acid versus lactone forms of statins except for fluvastatin were similar between rCYP2C9.1 and rCYP2C9.3. However, the degrees of inhibition by atorvastatin lactone, fluvastatin acid, fluvastatin lactone, lovastatin lactone, and pitavastatin lactone (Ki values) were significantly different between these variants. These results indicated that lactone forms of statins other than fluvastatin showed more potent inhibition of CYP2C9-catalyzed S-warfarin 7-hydroxylation than the corresponding acid forms. Furthermore, our results indicated that Ile359Leu substitution in CYP2C9 affected the inhibitory potencies of statins.
Sigma-Aldrich
7-Hydroxywarfarin
Supelco
Tenivastatin ammonium salt, analytical standard