Merck
  • Effects of a proinflammatory response on metabolic function of cultured, primary ruminal epithelial cells.
Effects of a proinflammatory response on metabolic function of cultured, primary ruminal epithelial cells.
Journal of dairy science (2020-11-03)
C Kent-Dennis, G B Penner
ABSTRACT
Inflammation of ruminal epithelium may occur during ruminal acidosis as a result of translocation and interaction of ruminal epithelial cells (REC) with molecules such as lipopolysaccharide (LPS). Such inflammation has been reported to alter cellular processes such as nutrient absorption, metabolic regulation, and energy substrate utilization in other cell types but has not been investigated for REC. The objectives of this study were to investigate the effects of LPS on metabolism of short-chain fatty acids by primary REC, as well as investigating the effects of media containing short-chain fatty acids on the proinflammatory response. Ruminal papillae from 9 yearling Speckle Park beef heifers were used to isolate and culture primary REC. Cells were grown in minimum essential medium (MEM) for 12 d before use and then reseeded in 24-well culture plates. The study was conducted as a 2 × 2 factorial, where cells were grown in unaltered MEM (REG) or medium containing 2 mM butyrate and 5 mM propionate (SCFA) with (50,000 EU/mL; +LPS) or without LPS (-LPS) for 24 h. Supernatant samples were collected for analysis of glucose and SCFA consumption. Cells were collected to determine the expression of mRNA for genes associated with inflammation (TNF, IL1B, CXCL2, CXCL8, PTGS2, and TLR4), purinergic signaling (P2RX7, ADORAB2, and CD73), nutrient transport [SLC16A1 (MCT1), SLC16A3 (MCT4), SLC5A8, and MCU], and cell metabolism [ACAT1, SLC2A1 (GLUT1), IGFBP3, and IGFBP5]. Protein expression of TLR4 and ketogenic enzymes (BDH1 and HMGCS1) were also analyzed using flow cytometry. Statistical analysis was conducted with the MIXED model of SAS version 9.4 (SAS Institute Inc., Cary, NC) with medium, LPS exposure, and medium × LPS interaction as fixed effects and animal within plate as a random effect. Cells tended to consume more glucose when exposed to LPS as opposed to no LPS exposure (31.8 vs. 28.7 ± 2.7), but consumption of propionate and butyrate was not influenced by LPS. Expression of TNF and IL1B was upregulated when exposed to LPS, and expression of CXCL2 and CXCL8 increased following LPS exposure with SCFA (medium × LPS). For cells exposed to LPS, we found a downregulation of ACAT1 and IGFBP5 and an upregulation of SLC2A1, SLC16A3, MCU, and IGFBP3. Medium with SCFA led to greater expression of MCU. SLC16A1 was upregulated in cells incubated with SCFA and without LPS compared with the other groups. Protein expression of ketogenic enzymes was not affected; however, BDH1 mean fluorescence intensity (MFI) expression tended to be less in cells exposed to LPS. These data are interpreted to indicate that when REC are exposed to LPS, they may increase glucose metabolism. Moreover, transport of solutes was affected by SCFA in the medium and by exposure to LPS. Overall, the results suggest that metabolic function of REC in vitro is altered by a proinflammatory response, which may lead to a greater glucose requirement.
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Medium 199, With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture