Merck
  • 1,2,5,6-Tetrakis(guanidino)-Naphthalenes: Electron Donors, Fluorescent Probes and Redox-Active Ligands.
1,2,5,6-Tetrakis(guanidino)-Naphthalenes: Electron Donors, Fluorescent Probes and Redox-Active Ligands.
Chemistry (Weinheim an der Bergstrasse, Germany) (2020-02-06)
Lukas Lohmeyer, Elisabeth Kaifer, Hubert Wadepohl, Hans-Jörg Himmel
ABSTRACT
New redox-active 1,2,5,6-tetrakis(guanidino)-naphthalene compounds, isolable and storable in the neutral and deep-green dicationic redox states and oxidisable further in two one-electron steps to the tetracations, are reported. Protonation switches on blue fluorescence, with the fluorescence intensity (quantum yield) increasing with the degree of protonation. Reactions with N-halogenosuccinimides or N-halogenophthalimides led to a series of new redox-active halogeno- and succinimido-/phthalimido-substituted derivatives. These highly selective reactions are proposed to proceed via the tri- or tetracationic state as the intermediate. The derivatives are oxidised reversibly at slightly higher potentials than that of the unsubstituted compounds to dications and further to tri- and tetracations. The integration of redox-active ligands in the transition-metal complexes shifts the redox potentials to higher values and also allows reversible oxidation in two potentially separated one-electron steps.
Sigma-Aldrich
Benzophenone imine, 95%
Sigma-Aldrich
2,6-Dibromonaphthalene, 97%