Merck
  • Synthesis and Properties of Cyclopentyl Cardo-Type Polyimides Based on Dicyclopentadiene.
Synthesis and Properties of Cyclopentyl Cardo-Type Polyimides Based on Dicyclopentadiene.
Polymers (2019-12-11)
Shih-Chieh Yeh, Jen-Yu Lee, Chung-Ta Hsieh, Ya-Chin Huang, Kuan-Syun Wang, Chien-Hsin Wu, Chien-Chieh Hu, Shu-Chen Chiang, Ru-Jong Jeng
ABSTRACT
A crucial polymer intermediate, 4-[1-(4-hydroxyphenyl)cyclopentyl]-phenol (bisphenol CP), was developed from dicyclopentadiene (DCPD), a key byproduct of the C5 fraction in petrochemicals. On the basis of bisphenol CP, a diamine, 4,4'-((cyclopentane-1,1-diylbis(4,1-phenylene))bis(oxy))-dianiline (cyclopentyl diamine; CPDA) was subsequently obtained through a nucleophilic substitution of bisphenol CP, followed by the hydrogenation process. By using the CPDA diamine, a series of polyimides with cyclopentyl (cardo) units on the backbone were prepared along with a reference polyimide (API-6F) based on 4,4'-(4,4'-(propane-2,2-diyl)bis(4,1-phenylene))bis(oxy)dianiline (BPAA), and 4,4'-(hexafluoroisopropylidene)-diphthalic anhydride (6FDA) for the exploration of structure-properties relationship. Thanks to the presence of cyclopentyl units, this type of cardo polyimides exhibited comparable tensile properties, especially a large elongation (25.4%). It is also worth noting that CPI-6F exhibited better solubility in organic solvents, such as NMP, DMAc, THF, and chloroform, than the other PIs. Gas separation properties were also evaluated for these cardo-type polyimides.
Sigma-Aldrich
Pyromellitic dianhydride, 97%
Sigma-Aldrich
Bis(benzonitrile)palladium(II) chloride, 95%