Merck
  • SIK inhibition in human myeloid cells modulates TLR and IL-1R signaling and induces an anti-inflammatory phenotype.
SIK inhibition in human myeloid cells modulates TLR and IL-1R signaling and induces an anti-inflammatory phenotype.
Journal of leukocyte biology (2015-11-22)
Maria Stella Lombardi, Corine Gilliéron, Damien Dietrich, Cem Gabay
ABSTRACT
Macrophage polarization into a phenotype producing high levels of anti-inflammatory IL-10 and low levels of proinflammatory IL-12 and TNF-α cytokines plays a pivotal role in the resolution of inflammation. Salt-inducible kinases synergize with TLR signaling to restrict the formation of these macrophages. The expression and function of salt-inducible kinase in primary human myeloid cells are poorly characterized. Here, we demonstrated that the differentiation from peripheral blood monocytes to macrophages or dendritic cells induced a marked up-regulation of salt-inducible kinase protein expression. With the use of 2 structurally unrelated, selective salt-inducible kinase inhibitors, HG-9-91-01 and ARN-3236, we showed that salt-inducible kinase inhibition significantly decreased proinflammatory cytokines (TNF-α, IL-6, IL-1β, and IL-12p40) and increased IL-10 secretion by human myeloid cells stimulated with TLR2 and-4 agonists. Differently than in mouse cells, salt-inducible kinase inhibition did not enhance IL-1Ra production in human macrophages. Salt-inducible kinase inhibition blocked several markers of proinflammatory (LPS + IFN-γ)-polarized macrophages [M(LPS + IFN-γ)] and induced a phenotype characterized by low TNF-α/IL-6/IL-12p70 and high IL-10. The downstream effects observed with salt-inducible kinase inhibitors on cytokine modulation correlated with direct salt-inducible kinase target (CREB-regulated transcription coactivator 3 and histone deacetylase 4) dephosphorylation in these cells. More importantly, we showed for the first time that salt-inducible kinase inhibition decreases proinflammatory cytokines in human myeloid cells upon IL-1R stimulation. Altogether, our results expand the potential therapeutic use of salt-inducible kinase inhibitors in immune-mediated inflammatory diseases.
Sigma-Aldrich
ARN-3236, ≥98% (HPLC)
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse