Merck
  • TRIAP1/PRELI complexes prevent apoptosis by mediating intramitochondrial transport of phosphatidic acid.
TRIAP1/PRELI complexes prevent apoptosis by mediating intramitochondrial transport of phosphatidic acid.
Cell metabolism (2013-08-13)
Christoph Potting, Takashi Tatsuta, Tim König, Mathias Haag, Timothy Wai, Mari J Aaltonen, Thomas Langer
ABSTRACT
Cardiolipin (CL), a mitochondria-specific glycerophospholipid, is required for diverse mitochondrial processes and orchestrates the function of various death-inducing proteins during apoptosis. Here, we identify a complex of the p53-regulated protein TRIAP1 (p53CSV) and PRELI in the mitochondrial intermembrane space (IMS), which ensures the accumulation of CL in mitochondria. TRIAP1/PRELI complexes exert lipid transfer activity in vitro and supply phosphatidic acid (PA) for CL synthesis in the inner membrane. Loss of TRIAP1 or PRELI impairs the accumulation of CL, facilitates the release of cytochrome c, and renders cells vulnerable to apoptosis upon intrinsic and extrinsic stimulation. Survival of TRIAP1- and PRELI-deficient cells is conferred by an excess of exogenously provided phosphatidylglycerol. Our results reveal a p53-dependent cell-survival pathway and highlight the importance of the CL content of mitochondrial membranes in apoptosis.
Product Description
Avanti
17:0-20:4 PG, Avanti Research - A Croda Brand
Sigma-Aldrich
GeneJuice® Transfection Reagent, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.