
Wogonoside

Synonym(s):
5,7-Dihydroxy 8-methoxyflavone 7-glucuronide, Glychionide B, Oroxindin, Wogonin 7-glucuronide
Empirical Formula (Hill Notation):
C22H20O11
CAS Number:
Molecular Weight:
460.39
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing