Skip to Content
Merck
Ulipristal acetate United States Pharmacopeia (USP) Reference Standard

Ulipristal acetate

Ulipristal acetate United States Pharmacopeia (USP) Reference Standard
Synonym(s):
(11β)-17-(Acetyloxy)-11-[4-(dimethylamino)phenyl]-19-norpregna-4,9-diene-3,20-dione, CDB-2914
Empirical Formula (Hill Notation):
C30H37NO4
CAS Number:
Molecular Weight:
475.62

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2