
Tetraammineplatinum(II) chloride hydrate

Linear Formula:
Pt(NH3)4Cl2·xH2O
CAS Number:
Molecular Weight:
334.11 (anhydrous basis)
Products
Brand
Formula Weight
Purity
Chemical Composition
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing