
Tanshinone I

Synonym(s):
Tanshinon I, Tanshinone A, Tanshinquinone I
Empirical Formula (Hill Notation):
C18H12O3
CAS Number:
Molecular Weight:
276.29
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing