
Sulpiride impurity A

Synonym(s):
2-(Aminomethyl)-1-ethylpyrrolidine
Empirical Formula (Hill Notation):
C7H16N2
CAS Number:
Molecular Weight:
128.22
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing