
Plastic additive 13

Synonym(s):
Tris(3,5-di-tert-butyl-4-hydroxybenzyl) isocyanurate, 1,3,5-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)-s-triazine-2,4,6(1H,3H,5H)-trione
Empirical Formula (Hill Notation):
C48H69N3O6
CAS Number:
Molecular Weight:
784.08
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing