Skip to Content
Merck
N,N,N′,N′-Tetramethylguanidine ≥99.0% (GC)

N,N,N′,N′-Tetramethylguanidine

N,N,N′,N′-Tetramethylguanidine ≥99.0% (GC)
Synonym(s):
1,1,3,3-Tetramethylguanidine, N,N,N′,N′-Tetramethylguanidine
Linear Formula:
[(CH3)2N]2C=NH
CAS Number:
Molecular Weight:
115.18

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing