Skip to Content
Merck
Naphthol AS phosphate >99% (TLC), histochemical substrate

Naphthol AS phosphate

Naphthol AS phosphate >99% (TLC), histochemical substrate
Synonym(s):
3-Hydroxy-2-naphthanilide phosphate
Empirical Formula (Hill Notation):
C17H14NO5P
CAS Number:
Molecular Weight:
343.27
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing