
Ibuprofen Impurity C

Synonym(s):
(2RS)-2-(4-Isobutylphenyl)propionamide, α-Methyl-4-(2-methylpropyl)benzeneacetamide, (2RS)-2-[4-(2-Methylpropyl)phenyl]propanamide, (±)-Ibuprofen amide
Empirical Formula (Hill Notation):
C13H19NO
CAS Number:
Molecular Weight:
205.30
Products
Brand
Formula Weight
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing