
Hydroxyprogesterone caproate

Synonym(s):
17α-Hydroxyprogesterone hexanoate, 17α-Hydroxyprogesterone caproate
Empirical Formula (Hill Notation):
C27H40O4
CAS Number:
Molecular Weight:
428.60
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2