
Fenamiphos-sulfoxide

Empirical Formula (Hill Notation):
C13H22NO4PS
CAS Number:
Molecular Weight:
319.36
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2