Ethyl 4,4,4-trifluoro-2-butynoate
Linear Formula:
CF3C≡CCO2C2H5
CAS Number:
Molecular Weight:
166.10
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing