
Dimetilan

Synonym(s):
1-(Dimethylcarbamoyl)-5-methyl-1H-pyrazol-3-yl dimethylcarbamate
Empirical Formula (Hill Notation):
C10H16N4O3
CAS Number:
Molecular Weight:
240.26
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing