Skip to Content
Merck
Dibromoacetic acid analytical standard

Dibromoacetic acid

Dibromoacetic acid analytical standard
Linear Formula:
Br2CHCOOH
CAS Number:
Molecular Weight:
217.84
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing