Skip to Content
Merck
4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) solution

4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Synonym(s):
4-(Methylnitrosoamino)-1-(3-pyridinyl)-1-butanone, 4-(Methylnitrosamino)-1-(3-pyridinyl)-1-butanone, NNK
Empirical Formula (Hill Notation):
C10H13N3O2
CAS Number:
Molecular Weight:
207.23

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique