Skip to Content
Merck
3-Phenylpyrazin-2-ol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

3-Phenylpyrazin-2-ol

3-Phenylpyrazin-2-ol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
3-Phenyl-1H-pyrazin-2-one
Empirical Formula (Hill Notation):
C10H8N2O
CAS Number:
Molecular Weight:
172.18

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade