Skip to Content
Merck
All Photos(1)

Key Documents

PZ0148

Sigma-Aldrich

CP-802079 hydrochloride hydrate

≥98% (HPLC)

Synonym(s):

({3-(4-Chlorophenyl)-3-[4-(thiazole-carbonyl)-phenoxy]-propyl}-methyl-amino)-acetic acid hydrochloride hydrate, N-[3-(4-Chlorophenyl)-3-[4-(2-thiazolylcarbonyl)phenoxy]propyl]-N-methyl-glycine hydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H21ClN2O4S · xHCl · yH2O
CAS Number:
Molecular Weight:
444.93 (anhydrous free base basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

very light brown

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

O.Cl.CN(CCC(Oc1ccc(cc1)C(=O)c2nccs2)c3ccc(Cl)cc3)CC(O)=O

InChI

1S/C22H21ClN2O4S.ClH.H2O/c1-25(14-20(26)27)12-10-19(15-2-6-17(23)7-3-15)29-18-8-4-16(5-9-18)21(28)22-24-11-13-30-22;;/h2-9,11,13,19H,10,12,14H2,1H3,(H,26,27);1H;1H2

InChI key

CVQXAUBKPDYXHF-UHFFFAOYSA-N

Biochem/physiol Actions

CP-802079 is a potent and selective glycine transporter type 1 (GlyT1) antagonist. Antagonists of GlyT1 increase levels of glycine in the synaptic cleft and, like direct glycine site agonists, they can augment NMDAR currents and NMDAR-mediated functions. CP-802,079 significantly increases the amplitude of the NMDAR currents and LTP.

Features and Benefits

This compound is featured on the Glycine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service