Skip to Content
Merck
All Photos(1)

Key Documents

8.06206

Sigma-Aldrich

1,8-Naphthalenediamine

for synthesis

Synonym(s):

1,8-Naphthalenediamine, 1,8-Diaminonaphthalene, 1,8-Naphthylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2
CAS Number:
Molecular Weight:
158.20
MDL number:
UNSPSC Code:
12352116
EC Index Number:
207-529-8
NACRES:
NA.22

Quality Level

form

powder

potency

800 mg/kg LD50, oral (Rat)

bp

205 °C/15.6 hPa

mp

61-65 °C

transition temp

flash point 195 °C

storage temp.

2-30°C

InChI

1S/C10H10N2/c11-8-5-1-3-7-4-2-6-9(12)10(7)8/h1-6H,11-12H2

InChI key

YFOOEYJGMMJJLS-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 97,0 %(m)
Melting range (lower value): ≥ 61 °C
Melting range (upper value): ≤ 65 °C
Identity (IR): conforms

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

392.0 °F - closed cup

Flash Point(C)

> 200 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service