Skip to Content
Merck
All Photos(1)

Key Documents

09-069

Sigma-Aldrich

Anti-ATF6 Antibody

Upstate®, from rabbit

Synonym(s):

Anti-ACHM7, Anti-ATF6A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ATF6(22926)

General description

ATF6 (Activating Transcription Factor 6, Cyclic AMP-dependent transcription factor ATF6) is a stress activated leucine zipper transcription factor involved in activating transcription of endoplasmic reticulum (ER) molecules in response to accumulation of unfolded proteins in the ER. ATF6 is originally a 90 kDa type II transmembrane glycoprotein that is converted to a 50 kDa soluble nuclear form, retaining its N-terminal domain, including the leucine zipper. Unfolded protein accumulation in the ER results in the cleavage of ATF6 to release its cytoplasmic domain that then enters the nucleus. ATF6 binds to an element designated ER Stress Response Element (ERSA) that appears in the promoter of glucose-regulated proteins (GRPs) such as GRP78, GPR94, and calreticulin.

Immunogen

KLH-tagged synthestic peptide synthesized to the N-terminal region of human ATF6 designated as the Cytoplasmic Domain.

Application

Detect ATF6 using this Anti-ATF6 Antibody validated for use in WB.
Research Category
Signaling

Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Routinely evaluated by western blot on HeLa lysate.

Target description

90 kDa

Physical form

Affinity purified
Affinity purified rabbit polyclonal IgG in storage buffer containing PBS with 0.05% sodium azide.

Storage and Stability

1 year at -20°C

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raphaël Santinelli et al.
Cells, 13(2) (2024-01-22)
Cystic Fibrosis (CF) is present due to mutations in the Cystic Fibrosis Transmembrane Conductance Regulator (CFTR) gene, the most frequent variant being p.phe508del. The CFTR protein is a chloride (Cl-) channel which is defective and almost absent of cell membranes
Benjamin L Farah et al.
Molecular genetics and metabolism, 122(3), 95-98 (2017-09-11)
GSD Ia (von Gierke Disease, Glycogen Storage Disease Type Ia) is a devastating genetic disorder with long-term sequelae, such as non-alcoholic fatty liver disease and renal failure. Down-regulated autophagy is involved in the development of hepatic metabolic dysfunction in GSD
Michael J Rigby et al.
Communications biology, 4(1), 454-454 (2021-04-14)
Nε-lysine acetylation in the ER lumen is a recently discovered quality control mechanism that ensures proteostasis within the secretory pathway. The acetyltransferase reaction is carried out by two type-II membrane proteins, ATase1/NAT8B and ATase2/NAT8. Prior studies have shown that reducing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service