Skip to Content
Merck
All Photos(1)

Key Documents

ALD00438

Sigma-Aldrich

Sodium (4-methoxyphenyl)methanesulfinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NaO3S
Molecular Weight:
208.21
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

reaction type: C-C Bond Formation

mp

260-265 °C

SMILES string

[O-]S(CC1=CC=C(OC)C=C1)=O.[Na+]

InChI

1S/C8H10O3S.Na/c1-11-8-4-2-7(3-5-8)6-12(9)10;/h2-5H,6H2,1H3,(H,9,10);/q;+1/p-1

InChI key

OKSPESPSBLOXDC-UHFFFAOYSA-M

Application

The following Baran Sulfinate is a part of a toolbox of diversification reagents; which are ideal for the late-stage functionalization of nitrogen-containing heterocycles. The regioselectivity can be effectively tuned by modification of the pH and solvent selection.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service