Merck
  • Immunohistochemical and serological characterization of membranous nephropathy in children and adolescents.
Immunohistochemical and serological characterization of membranous nephropathy in children and adolescents.
Pediatric nephrology (Berlin, Germany) (2017-10-17)
Anne K Dettmar, Thorsten Wiech, Markus J Kemper, Armin Soave, Michael Rink, Jun Oh, Rolf A K Stahl, Elion Hoxha
ABSTRACT
Membranous nephropathy (MN) is a common cause of nephrotic syndrome in adults, but is less frequent in children. Antibodies against four antigens leading to MN have been described in children: phospholipase A Twelve children with MN were included in this study. Sera of all patients were analyzed for antibodies against PLA Six patients were PLA In children with MN, PLA
Product Description
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified, viscosity 4599 mPa.s (25 °C)
Sigma-Aldrich
1-Ethyl-2-pyrrolidone, 98%