- MK-8776, a novel Chk1 inhibitor, exhibits an improved radiosensitizing effect compared to UCN-01 by exacerbating radiation-induced aberrant mitosis.
MK-8776, a novel Chk1 inhibitor, exhibits an improved radiosensitizing effect compared to UCN-01 by exacerbating radiation-induced aberrant mitosis.
Translational oncology (2017-05-28)
Motofumi Suzuki, Tohru Yamamori, Tomoki Bo, Yuri Sakai, Osamu Inanami
PMID28550769
ABSTRACT
Checkpoint kinase 1 (Chk1) is an evolutionarily conserved serine/threonine kinase that plays an important role in G