Merck
  • Mesothelin/mucin 16 signaling in activated portal fibroblasts regulates cholestatic liver fibrosis.
Mesothelin/mucin 16 signaling in activated portal fibroblasts regulates cholestatic liver fibrosis.
The Journal of clinical investigation (2017-03-14)
Yukinori Koyama, Ping Wang, Shuang Liang, Keiko Iwaisako, Xiao Liu, Jun Xu, Mingjun Zhang, Mengxi Sun, Min Cong, Daniel Karin, Kojiro Taura, Chris Benner, Sven Heinz, Tapan Bera, David A Brenner, Tatiana Kisseleva
ABSTRACT
Cholestatic liver fibrosis is caused by obstruction of the biliary tract and is associated with early activation of portal fibroblasts (PFs) that express Thy-1, fibulin 2, and the recently identified marker mesothelin (MSLN). Here, we have demonstrated that activated PFs (aPFs) and myofibroblasts play a critical role in the pathogenesis of liver fibrosis induced by bile duct ligation (BDL). Conditional ablation of MSLN+ aPFs in BDL-injured mice attenuated liver fibrosis by approximately 50%. Similar results were observed in MSLN-deficient mice (Msln-/- mice) or mice deficient in the MSLN ligand mucin 16 (Muc16-/- mice). In vitro analysis revealed that MSLN regulates TGF-β1-inducible activation of WT PFs by disrupting the formation of an inhibitory Thy-1-TGFβRI complex. MSLN also facilitated the FGF-mediated proliferation of WT aPFs. Therapeutic administration of anti-MSLN-blocking Abs attenuated BDL-induced fibrosis in WT mice. Liver specimens from patients with cholestatic liver fibrosis had increased numbers of MSLN+ aPFs/myofibroblasts, suggesting that MSLN may be a potential target for antifibrotic therapy.
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
Anti-MSLN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution