Merck
  • Identification of 4-Trimethylaminobutyraldehyde Dehydrogenase (TMABA-DH) as a Candidate Serum Autoantibody Target for Kawasaki Disease.
Identification of 4-Trimethylaminobutyraldehyde Dehydrogenase (TMABA-DH) as a Candidate Serum Autoantibody Target for Kawasaki Disease.
PloS one (2015-05-27)
Atsuko Matsunaga, Yutaka Harita, Yoshio Shibagaki, Nobutaka Shimizu, Kazuhiko Shibuya, Hiroshi Ono, Hitoshi Kato, Takashi Sekine, Naoko Sakamoto, Takashi Igarashi, Seisuke Hattori
ABSTRACT
Kawasaki disease (KD), an acute vasculitis that preferentially affects coronary arteries, is still the leading cause of acquired heart disease in children. Although the involvement of immune system malfunction in the onset of KD is suggested, its etiology still remains to be clarified. We investigated autoantibodies in KD patients, which are frequently found in sera from patients with autoimmune diseases, vasculitides and arteritides. We performed two-dimensional western blotting and LC-MS/MS to analyze the antigens of autoantibodies, detected two protein spots with 4 out of 24 sera from KD patients but not with 6 control sera, and identified the antigens as 4-trimethylaminobutyraldehyde dehydrogenase (TMABA-DH). A slot blot analysis with TMABA-DH as an antigen also revealed higher reactivities of patients' sera than control sera (positive rates: 18/43 vs 3/41). Using an enzyme-linked immunosorbent assay (ELISA), we found that the reactivity of anti-TMABA-DH antibodies in sera from KD patients was significantly higher than that in sera from age-matched controls. The optimal cut-off value of 0.043 had a sensitivity of 83.7% and a specificity of 80.0% in detecting KD patients (positive rates: 37/43 for KD patients, 9/41 for controls). Immunohistochemistry performed on thin sections of rat heart revealed that TMABA-DH colocalized with myosin light chains in cardiac myocytes. Patient sera with high reactivity gave similar immunostaining pattern. These results suggest that the detection of anti-TMABA-DH autoantibody could be a potential strategy for a diagnosis of KD.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Anti-ALDH9A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Ultrapure Acetonitrile