Merck
  • Indolosesquiterpene alkaloids from the Cameroonian medicinal plant Polyalthia oliveri (Annonaceae).
Indolosesquiterpene alkaloids from the Cameroonian medicinal plant Polyalthia oliveri (Annonaceae).
Phytochemistry (2014-07-21)
Simeon Fogue Kouam, Alain Wembe Ngouonpe, Marc Lamshöft, Ferdinand Mouafo Talontsi, Jonathan O Bauer, Carsten Strohmann, Bonaventure Tchaleu Ngadjui, Hartmut Laatsch, Michael Spiteller
ABSTRACT
The stem bark of Polyalthia oliveri was screened for its chemical constituents using liquid chromatography high resolution mass spectrometry resulting in the isolation of three indolosesquiterpene alkaloids named 8α-polyveolinone (1), N-acetyl-8α-polyveolinone (2) and N-acetyl-polyveoline (3), together with three known compounds, dehydro-O-methylisopiline (4), N-methylurabaine (5) and polycarpol (6). The structures of the compounds were elucidated by means of high resolution mass spectrometry and different NMR techniques and chemical transformations. Their absolute configurations were assigned by ab-initio calculation of CD and ORD data (for 2 and 3) and X-ray diffraction analysis (for 2). Compounds 2 and 3 exhibited moderate antiplasmodial activity against erythrocytic stages of chloroquine-sensitive Plasmodium falciparum NF54 strain and low cytotoxicity on rat skeletal myoblast (L6) cell line.
Product Description
Pyrimethamine, European Pharmacopoeia (EP) Reference Standard
Supelco
Pyrimethamine, VETRANAL®, analytical standard
USP
Pyrimethamine, United States Pharmacopeia (USP) Reference Standard