Merck
  • Population pharmacokinetics of fusidic acid: rationale for front-loaded dosing regimens due to autoinhibition of clearance.
Population pharmacokinetics of fusidic acid: rationale for front-loaded dosing regimens due to autoinhibition of clearance.
Antimicrobial agents and chemotherapy (2012-11-14)
Jürgen B Bulitta, Olanrewaju O Okusanya, Alan Forrest, Sujata M Bhavnani, Kay Clark, J Gordon Still, Prabhavathi Fernandes, Paul G Ambrose
ABSTRACT
The objectives of this analysis were to develop a population pharmacokinetic (PK) model to describe the absorption and disposition of fusidic acid after single and multiple doses and to determine the effect of food on the rate and extent of bioavailability. Plasma PK data from three phase 1 studies (n = 75; n = 14 with and without food) in which healthy subjects received sodium fusidate (500 to 2,200 mg) as single or multiple oral doses every 8 h (q8h) or q12h for up to 7 days were modeled using S-ADAPT (MCPEM algorithm). Accumulation of fusidic acid after multiple doses was more than that predicted based on single-dose data. The PK of fusidic acid was best described using a time-dependent mixed-order absorption process, two disposition compartments, and a turnover process to describe the autoinhibition of clearance. The mean total clearance (% coefficient of variation) was 1.28 liters/h (33%) and the maximum extent of autoinhibition was 71.0%, with a 50% inhibitory concentration (IC(50)) of 46.3 mg/liter (36%). Food decreased the extent of bioavailability by 18%. As a result of the autoinhibition of clearance, steady state can be achieved earlier with dosing regimens that contain higher doses (after 8 days for 750 mg q12h and 1 day for 1,500 mg q12h on day 1 followed by 600 mg q12h versus 3 weeks for 500 mg q12h). Given that large initial doses autoinhibit the clearance of fusidic acid, this characteristic provides a basis for the administration of front-loaded dosing regimens of sodium fusidate which would allow for effective concentrations to be achieved early in therapy.
Sigma-Aldrich
Fusidic acid sodium salt, ≥98% (TLC)
Sigma-Aldrich
Fusidic acid