Merck
  • The use of oxyhalogen in photocatalytic reaction to remove o-chloroaniline in TiO2 dispersion.
The use of oxyhalogen in photocatalytic reaction to remove o-chloroaniline in TiO2 dispersion.
Chemosphere (2006-11-23)
W K Choy, W Chu
ABSTRACT
Photodecay of o-chloroaniline (o-ClA) in various combinations of UV sources, TiO2, and oxyhalogens was investigated. To improve the conventional photocatalytic process by using UV/TiO2, the addition of oxyhalogens (ClO3(-), BrO3(-) and IO3(-)) into UV/TiO2 system was studied and the effect in such addition is very encouraging for all the selected additives. Oxyhalogens are capable of deferring the electron-hole recombination of TiO2 which significantly improved its catalytic performance. The presence of IO3(-) in UV/TiO2 resulted in the fastest o-ClA decay among three oxhalogens at the same dosage. The decay of o-ClA in UV/TiO2/oxyhalogen process is characterized by a two-stage pseudo-kinetics, where a faster initial decay was followed by a retardation state. A mathematics model was successfully established for the prediction of the two-stage decay of o-ClA in UV/TiO2/IO3(-) with any designed [IO3(-)] concentration.
Sigma-Aldrich
2-Chloroaniline, ≥99.5% (GC)
Sigma-Aldrich
2-Chloroaniline, technical, ≥98.0% (GC)