Merck
  • Semiconductor-mediated photocatalysed degradation of two selected azo dye derivatives, amaranth and bismarck brown in aqueous suspension.
Semiconductor-mediated photocatalysed degradation of two selected azo dye derivatives, amaranth and bismarck brown in aqueous suspension.
Journal of hazardous materials (2005-08-13)
M Abu Tariq, M Faisal, M Muneer
ABSTRACT
Semiconductor-mediated photocatalysed degradation of two selected azo dye derivatives such as amaranth (1) and bismarck brown (2) has been investigated in aqueous suspension by monitoring the change in substrate concentration employing UV spectroscopic analysis technique as a function of irradiation time. The degradation was studied under different conditions such as types of TiO(2), pH, substrate concentration, catalyst concentration, and in the presence of electron acceptors such as hydrogen peroxide (H(2)O(2)), potassium bromate (KBrO(3)) and ammonium persulphate (NH(4))(2)S(2)O(8) besides air. The degradation rates were found to be strongly influenced by all the above parameters. The photocatalyst Degussa P25 showed comparatively highest photocatalytic activity. The dye derivative, bismarck brown (2) was found to degrade faster than amaranth dye (1).
Sigma-Aldrich
Bismarck Brown Y, certified by the BSC, Dye content 50 %