Merck
  • Pyrophosphatase overexpression is associated with cell migration, invasion, and poor prognosis in gastric cancer.
Pyrophosphatase overexpression is associated with cell migration, invasion, and poor prognosis in gastric cancer.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2012-07-17)
Sang-Ho Jeong, Gyung Hyuck Ko, Young Hyun Cho, Young-Joon Lee, Bok-Im Cho, Woo-Song Ha, Sang-Kyung Choi, Jae Won Kim, Chang Won Lee, Yoon Seok Heo, Seok Hwan Shin, Jiyun Yoo, Soon-Chan Hong
ABSTRACT
Inorganic pyrophosphatase (PPase) catalyzes the hydrolysis of pyrophosphate to form orthophosphate. Pyrophosphate can substitute for ATP under certain circumstances. We previously conducted a proteomic analysis to investigate tumor-specific protein expression in gastric cancer, and PPase was identified as a potential gastric tumor-specific marker; it was therefore selected for further study. Clinicopathological analysis, using proteomic analysis and immunohistochemistry, was used to validate PPase as a prognostic marker in gastric cancers. Proteomic analysis showed that PPase was overexpressed in patients with lymph node (LN) metastases and high tumor node metastasis (TNM) stages (p < 0.05). Based on immunohistochemistry, patients whose tumors overexpressed PPase had higher T stages, LN metastasis, a higher TNM stage, a higher cancer recurrence rate, and shorter survival times than patients whose tumors exhibited PPase underexpression (p < 0.05). Gain-of-function and loss-of-function approaches were employed to examine the malignant phenotypes of PPase-overexpressing or PPase-depleted cells. A decrease in PPase expression caused a significant decrease in gastric cancer cell migration and invasion in vitro, whereas forced overexpression of PPase enhanced migration but not invasion. Our findings indicate that PPase is involved in gastric tumor progression and that PPase may be a useful marker for poor prognosis of human gastric cancers.
Sigma-Aldrich
Pyrophosphatase, Inorganic from Escherichia coli, recombinant, expressed in E. coli, lyophilized powder, ≥90%, ≥800 units/mg protein
Sigma-Aldrich
Pyrophosphatase, Inorganic from baker′s yeast (S. cerevisiae), powder, ≥500 units/mg protein (E1%/280)