Merck
  • Altered Distribution and Expression of Syndecan-1 and -4 as an Additional Hallmark in Psoriasis.
Altered Distribution and Expression of Syndecan-1 and -4 as an Additional Hallmark in Psoriasis.
International journal of molecular sciences (2022-06-25)
Eleni Koliakou, Manthou Maria Eleni, Ioanna Koumentakou, Nikolaos Bikiaris, Polyanthi Konstantinidou, Patricia Rousselle, Doxakis Anestakis, Elisabeth Lazaridou, Evangelia Kalloniati, Dimosthenis Miliaras, Anna Michopoulou
ABSTRACT
Syndecans act as independent co-receptors to exert biological activities and their altered function is associated with many pathophysiological conditions. Here, syndecan-1 and -4 were examined in lesional skin of patients with psoriasis. Immunohistochemical staining confirmed altered syndecan-1 distribution and revealed absence of syndecan-4 expression in the epidermis. Fibronectin (FN)-known to influence inflammation and keratinocyte hyperproliferation via α5β1 integrin in psoriasis-was also decreased. Syndecan-1 and -4 expression was analyzed in freshly isolated lesional psoriatic human keratinocytes (PHK) characterized based on their proliferation and differentiation properties. mRNA levels of syndecan-1 were similar between healthy and PHK, while syndecan-4 was significantly decreased. Cell growth and release of the pro-inflammatory Tumor Necrosis Factor-alpha (TNFα) were selectively and significantly induced in PHKs plated on FN. Results from co-culture of healthy keratinocytes and psoriatic fibroblasts led to the speculation that at least one factor released by fibroblasts down-regulate syndecan-1 expression in PHK plated on FN. To assay if biological treatments for psoriasis target keratinocyte proliferation, gelatin-based patches enriched with inteleukin (IL)-17α or TNFα blockers were prepared and tested using a full-thickness healthy epidermal model (Phenion®). Immunohistochemistry analysis showed that both blockers impacted the localisation of syndecan-1 within the refined epidermis. These results provide evidence that syndecans expression are modified in psoriasis, suggesting that they may represent markers of interest in this pathology.
Sigma-Aldrich
Human Plasma Fibronectin Purified Protein, from human plasma, liquid, 1 mg/mL (100 MG pack size is lyophilized), purified protein, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-TNF antibody produced in mouse, clone M1-C4, purified immunoglobulin, buffered aqueous solution