Merck
  • High intensity interval training promotes total and visceral fat mass loss in obese Zucker rats without modulating gut microbiota.
High intensity interval training promotes total and visceral fat mass loss in obese Zucker rats without modulating gut microbiota.
PloS one (2019-04-10)
Florie Maillard, Emilie Vazeille, Pierre Sauvanet, Pascal Sirvent, Lydie Combaret, Antoine Sourdrille, Vivien Chavanelle, Richard Bonnet, Yolanda Fernandez Otero, Geoffrey Delcros, Nicolas Barnich, Nathalie Boisseau
ABSTRACT
Increased visceral adipose tissue and dysbiosis in the overweight and obese promote chronic inflammation. The aim of this study was to compare the effects of moderate-intensity continuous training (MICT) and high-intensity interval training (HIIT) on the gut-adipose tissue cross-talk in obese Zucker rats. Obese male Zucker rats (n = 36) were divided in three groups: MICT (12m.min-1 for 51min), HIIT (6 sets at 18 m.min-1 for 4min followed by 3min at 10m.min-1) and controls (CONT; no exercise). The animals ran on a treadmill 5 days/week for 10 weeks. Body composition, glycaemic control, lipid profile, inflammation, lipolysis signalling in subcutaneous and visceral adipose tissue, intestinal permeability (tight junctions and plasma lipopolysaccharide binding protein; LBP), and gut microbiota composition were assessed in the three groups. After 10 weeks of exercise, total and epididymal fat mass decreased only in the HIIT group. The α/β adrenergic receptor RNA ratio in subcutaneous adipose tissue increased only in the HIIT group. The expression level of phosphorylated hormone-sensitive lipase was not modified by training. Both HIIT and MICT decreased inflammation (plasma myeloperoxidase and keratinocyte-derived chemokine secretion in adipose tissue) and improved glucose metabolism. Zonula occludens-1 and occludin were upregulated in the HIIT group. Plasma LBP was similarly reduced in both training groups. HIIT and MICT did not affect gut microbiota composition. In obese Zucker rats, HIIT and MICT improved inflammation and glucose metabolism. In contrast, only HIIT decreased total and visceral fat mass. These adaptations were not associated with modifications in gut microbiota composition.
Sigma-Aldrich
MTP Taq DNA Polymerase, Taq DNA Polymerase, free of DNA contaminants
Sigma-Aldrich
10X MTP Taq Buffer, 10X buffer, free of DNA contaminants; use with MTP Taq DNA Polymerase