Merck
  • Assessment of Sperm Binding Capacity in the Tubal Reservoir Using a Bovine Ex Vivo Oviduct Culture and Fluorescence Microscopy.
Assessment of Sperm Binding Capacity in the Tubal Reservoir Using a Bovine Ex Vivo Oviduct Culture and Fluorescence Microscopy.
Methods and protocols (2021-10-27)
Miguel Camara Pirez, Simeng Li, Sabine Koelle
ABSTRACT
Sperm binding within the oviductal sperm reservoir plays an important role for reproductive success by enabling sperm survival and maintaining fertilizing capacity. To date, numerous in vitro technologies have been established to measure sperm binding capacity to cultured oviductal cells or oviductal explants. However, these methods do not accurately represent the microenvironment and complex multi-molecular nature of the oviduct. In this paper, we describe a novel protocol for assessing sperm binding capacity in the tubal sperm reservoir using an ex vivo oviduct culture in the bovine model. This protocol includes the staining of frozen-thawed bovine spermatozoa with the DNA-binding dye Hoechst 33342, the co-incubation of stained sperm in closed segments of the oviduct and the visualization and quantification of bound spermatozoa by fluorescence microscopy. By generating overlays of multiple Z-stacks of randomly selected regions of interest (ROIs), spermatozoa bound in the sperm reservoir can be visualized and quantified within the 3D arrangement of the oviductal folds. This method, which is applicable to multiple species, can be used to assess individual sperm binding capacity in males for prognostic purposes as well as to assess the impact of diseases and medications on the formation of the sperm reservoir in the oviduct in humans and animals.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Microcentrifuge tubes with attached lid, capacity 1.5 mL