Merck
  • Eph-B4 mediates vein graft adaptation by regulation of endothelial nitric oxide synthase.
Eph-B4 mediates vein graft adaptation by regulation of endothelial nitric oxide synthase.
Journal of vascular surgery (2016-01-29)
Mo Wang, Michael J Collins, Trenton R Foster, Hualong Bai, Takuya Hashimoto, Jeans M Santana, Chang Shu, Alan Dardik
ABSTRACT
Vein graft adaptation is characterized by loss of expression of the tyrosine kinase receptor Eph-B4, the embryonic determinant of venous identity, without increased expression of its ligand ephrin-B2, the embryonic determinant of arterial identity. Endothelial nitric oxide synthase (eNOS) is an important mediator of vessel remodeling. We hypothesized that the mechanism of action of Eph-B4 during vein graft adaptation might be through regulation of downstream eNOS activity. Mouse lung endothelial cells were stimulated with ephrin-B2/Fc, without and with preclustering, without and with the eNOS inhibitor Nω-nitro-l-arginine methyl ester hydrochloride or the Eph-B4 inhibitor NVP-BHG712, and assessed by Western blot and immunofluorescence for eNOS and Eph-B4 phosphorylation. Nitric oxide (NO) production was assessed using an NO-specific chemiluminescence analyzer. Cell migration was assessed using a Transwell assay. Human and mouse vein graft specimens were examined for eNOS activity by Western blot, and vessel remodeling was assessed in vein grafts in wild-type or eNOS knockout mice. Ephrin-B2/Fc stimulated both Eph-B4 and eNOS phosphorylation in a bimodal temporal distribution (n = 4; P < .05), with preclustered ephrin-B2/Fc causing prolonged peak Eph-B4 and eNOS phosphorylation as well as altered subcellular localization (n = 4; P < .05). Ephrin-B2/Fc increased NO release (n = 3; P < .01) as well as increased endothelial cell migration (n = 6; P < .05) in an eNOS-dependent fashion. Both human and mouse vein grafts showed increased eNOS phosphorylation compared with normal veins (n = 3; P < .05). Vein grafts from eNOS knockout mice showed less dilation and less wall thickening compared with wild-type vein grafts (n = 7; P < .05). eNOS is a mediator of vein graft adaptation to the arterial environment. Eph-B4 stimulates eNOS phosphorylation in vitro and may mediate vein graft adaptation by regulation of eNOS activity in vivo.
Sigma-Aldrich
Nω-Nitro-L-arginine methyl ester hydrochloride, ≥97% (TLC), powder
Sigma-Aldrich
NVP-BHG712, ≥98% (HPLC)