Merck
  • Characterization of a novel, water-soluble hydrogen sulfide-releasing molecule (GYY4137): new insights into the biology of hydrogen sulfide.
Characterization of a novel, water-soluble hydrogen sulfide-releasing molecule (GYY4137): new insights into the biology of hydrogen sulfide.
Circulation (2008-04-30)
Ling Li, Matthew Whiteman, Yan Yi Guan, Kay Li Neo, Yvonne Cheng, Shiau Wei Lee, Yujun Zhao, Rajamanian Baskar, Choon-Hong Tan, Philip K Moore
ABSTRACT
The potential biological significance of hydrogen sulfide (H(2)S) has attracted growing interest in recent years. The aim of this study was to characterize a novel, water-soluble, slow-releasing H(2)S compound [morpholin-4-ium 4 methoxyphenyl(morpholino) phosphinodithioate (GYY4137)] and evaluate its use as a tool to investigate the cardiovascular biology of this gas. The acute vasorelaxant effect of drugs was assessed in rat aortic rings and perfused rat kidney in vitro and in the anesthetized rat in vivo. The chronic effect of GYY4137 on blood pressure in normotensive and spontaneously hypertensive rats was determined by tail-cuff plethysmography. GYY4137 released H(2)S slowly both in aqueous solution in vitro and after intravenous or intraperitoneal administration in anesthetized rats in vivo. GYY4137 caused a slow relaxation of rat aortic rings and dilated the perfused rat renal vasculature by opening vascular smooth muscle K(ATP) channels. GYY4137 did not affect rat heart rate or force of contraction in vitro. GYY4137 exhibited antihypertensive activity as evidenced by ability to reduce N(G)-nitro-L-arginine methyl ester-evoked hypertension in the anesthetized rat and after chronic (14-day) administration in spontaneously hypertensive rats. These results identify GYY4137 as a slow-releasing H(2)S compound with vasodilator and antihypertensive activity. GYY4137 is likely to prove useful in the study of the many and varied biological effects of H(2)S. GYY4137 may also prove of therapeutic value in cardiovascular disease.
Sigma-Aldrich
GYY4137 Dichloromethane complex, ≥98% (HPLC)